4-Azido-L-phenylalanine

Supplier: Apollo Scientific
Warning

Synonyms: H-4-Azido-Phe-OH

OR962222-100MG OR962222-1G OR962222-250MG
APOSOR962222-100MGEA 219 EUR
APOSOR962222-100MG APOSOR962222-1G APOSOR962222-250MG
4-Azido-L-phenylalanine
4-Azido-L-phenylalanine
Formula: C₉H₁₀N₄O₂
MW: 206,205 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00238100
CAS Number: 33173-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR