2-((4-Cyanophenyl)amino)acetic acid 95%

Supplier: Apollo Scientific

Synonyms: N-(4-CN-Phenyl)-Gly-OH, N-(4-Cyanophenyl)glycine

OR962439-25G OR962439-100G OR962439-500G
APOSOR962439-25GEA 46.6 EUR
APOSOR962439-25G APOSOR962439-100G APOSOR962439-500G
2-((4-Cyanophenyl)amino)acetic acid 95%
2-((4-Cyanophenyl)amino)acetic acid
Formula: C₉H₈N₂O₂
MW: 176,1700 g/mol
Melting Pt: 237 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01464022
CAS Number: 42288-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR