1-Propanesulphonic acid 95%
Supplier: Apollo Scientific
Synonyms:
Propane-1-sulphonic acid
OR962510-1G
APOSOR962510-1GEA
62.5
EUR
APOSOR962510-1G
1-Propanesulphonic acid 95%
1-Propanesulphonic acid
Formula:
C₃H₈O₃S MW: 124,16 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00041889 CAS Number: 5284-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...