(5-bromothien-2-yl)methanol 98%

Supplier: Apollo Scientific

Synonyms: (5-Bromothien-2-yl)methanol, 5-Bromo-2-thiophenemethanol

OR962641-1G OR962641-25G OR962641-5G
APOSOR962641-1GEA 65.5 EUR
APOSOR962641-1G APOSOR962641-25G APOSOR962641-5G
(5-bromothien-2-yl)methanol 98%
(5-bromothien-2-yl)methanol
Formula: C₅H₅BrOS
MW: 193,058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06202626
CAS Number: 79387-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR