N-(3-Methoxy-4-nitrophenyl)-N,N-dimethylamine 95%
Supplier: Apollo Scientific
OR962803-1G
OR962803-250MG
OR962803-5G
APOSOR962803-1GEA
234
EUR
APOSOR962803-1G
APOSOR962803-250MG
APOSOR962803-5G
N-(3-Methoxy-4-nitrophenyl)-N,N-dimethylamine 95%
N-(3-Methoxy-4-nitrophenyl)-N,N-dimethylamine
Formula:
C₉H₁₂N₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD09972264 CAS Number: 14703-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...