N-Boc-1,4-diaminobutane-HCl
Supplier: Apollo Scientific
|
Warning
|
OR963265-25G
OR963265-5G
APOSOR963265-25GEA
114
EUR
APOSOR963265-25G
APOSOR963265-5G
N-Boc-1,4-diaminobutane-HCl
N-Boc-1,4-diaminobutane-HCl
Formula:
C₉H₂₁ClN₂O₂ MW: 224,729 g/mol Melting Pt: 155…156 °C Storage Temperature: Ambient |
MDL Number:
MFCD02090802 CAS Number: 33545-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...