2-Isocyanatoethyl acrylate 95%

Supplier: Apollo Scientific

Synonyms: 2-(Acryloyloxy)ethyl Isocyanate, Acrylic Acid 2-Isocyanatoethyl Ester

OR963987-1G OR963987-25G OR963987-5G
APOSOR963987-1GEA 125 EUR
APOSOR963987-1G APOSOR963987-25G APOSOR963987-5G
2-Isocyanatoethyl acrylate 95%
2-Isocyanatoethyl acrylate

  • May form small amount of sediment.

Formula: C₆H₇NO₃
MW: 141,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11112219
CAS Number: 13641-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR