6-Methoxy-1,2,3,4-tetrahydroquinoline 95%

Supplier: Apollo Scientific

OR964112-5G
APOSOR964112-5GEA 143 EUR
APOSOR964112-5G
6-Methoxy-1,2,3,4-tetrahydroquinoline 95%
6-Methoxy-1,2,3,4-tetrahydroquinoline
Formula: C₁₀H₁₃NO
MW: 163,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00090494
CAS Number: 120-15-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR