N-Carbobenzoxy-D-threonine 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Z-D-Thr-OH, N-Cbz-D-threonine, (2R,3S)-2-(((Benzyloxy)carbonyl)amino)-3-hydroxybutanoic acid
OR964218-100G
OR964218-25G
APOSOR964218-100GEA
452
EUR
APOSOR964218-100G
APOSOR964218-25G
N-Carbobenzoxy-D-threonine 97%
N-Carbobenzoxy-D-threonine
Formula:
C₁₂H₁₅NO₅ MW: 253,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037808 CAS Number: 80384-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...