Methyl-3-cyanocinnamate 95%
Supplier: Apollo Scientific
Synonyms:
3-(3-Cyano-phenyl)-acrylic acid methyl ester
OR964221-1G
OR964221-250MG
APOSOR964221-1GEA
190
EUR
APOSOR964221-1G
APOSOR964221-250MG
Methyl-3-cyanocinnamate 95%
Methyl-3-cyanocinnamate
Formula:
C₁₁H₉NO₂ MW: 187,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08064268 CAS Number: 52116-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...