Boc-L-Cys-OH 97%

Supplier: Apollo Scientific

Synonyms: N-Boc-L-cysteine, (R)-2-((tert-Butoxycarbonyl)amino)-3-mercaptopropanoic acid

OR964365-1G OR964365-25G OR964365-5G
APOSOR964365-1GEA 34.8 EUR
APOSOR964365-1G APOSOR964365-25G APOSOR964365-5G
Boc-L-Cys-OH 97%
Boc-L-Cys-OH
Formula: C₈H₁₅NO₄S
MW: 221,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065565
CAS Number: 20887-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR