D-Phenylalaninamide hydrochloride 98%

Supplier: Apollo Scientific
Warning

Synonyms: H-D-Phe-NH2·HCl

OR964553-100G OR964553-25G OR964553-5G
APOSOR964553-100GEA 905 EUR
APOSOR964553-100G APOSOR964553-25G APOSOR964553-5G
D-Phenylalaninamide hydrochloride 98%
D-Phenylalaninamide hydrochloride
Formula: C₉H₁₂N₂O·HCl
Storage Temperature: Ambient
MDL Number: MFCD00153454
CAS Number: 71666-94-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR