5-Methoxy-7-azaindole 97%
Supplier: Apollo Scientific
Synonyms:
5-Methoxy-1H-pyrrolo[2,3-b]pyridine
OR964856-5G
APOSOR964856-5GEA
87
EUR
APOSOR964856-5G
5-Methoxy-7-azaindole 97%
5-Methoxy-7-azaindole
Formula:
C8H8N2O MW: 148,1600 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06659675 CAS Number: 183208-36-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...