3-Bromophthalic anhydride 97%
Supplier: Apollo Scientific
Synonyms:
4-Bromoisobenzofuran-1,3-dione
OR964928-1G
OR964928-25G
OR964928-5G
APOSOR964928-1GEA
50.9
EUR
APOSOR964928-1G
APOSOR964928-25G
APOSOR964928-5G
3-Bromophthalic anhydride 97%
3-Bromophthalic anhydride
Formula:
C₈H₃BrO₃ Storage Temperature: Ambient |
MDL Number:
MFCD09800402 CAS Number: 82-73-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...