3-Chloropropyl acetate 97%

Supplier: Apollo Scientific

OR964999-500G
APOSOR964999-500GEA 111 EUR
APOSOR964999-500G
3-Chloropropyl acetate 97%
3-Chloropropyl acetate
Formula: C₅H₉ClO₂
MW: 136,58 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001000
CAS Number: 628-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR