3-(Methylaminomethyl)benzonitrile 95%

Supplier: Apollo Scientific

OR965101-1G OR965101-250MG
APOSOR965101-1GEA 488 EUR
APOSOR965101-1G APOSOR965101-250MG
3-(Methylaminomethyl)benzonitrile 95%
3-(Methylaminomethyl)benzonitrile
Formula: C₉H₁₀N₂
MW: 146,193 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09731650
CAS Number: 90389-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR