4-Phenyl-1H-pyrazole 98%

Supplier: Apollo Scientific

OR965552-1G OR965552-250MG
APOSOR965552-1GEA 322 EUR
APOSOR965552-1G APOSOR965552-250MG
4-Phenyl-1H-pyrazole 98%
4-Phenyl-1H-pyrazole
Formula: C₉H₈N₂
Storage Temperature: Ambient
MDL Number: MFCD00462204
CAS Number: 10199-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR