2-Acetylquinoxaline 95%

Supplier: Apollo Scientific

OR965860-1G OR965860-5G
APOSOR965860-1GEA 125 EUR
APOSOR965860-1G APOSOR965860-5G
2-Acetylquinoxaline 95%
2-Acetylquinoxaline
Formula: C₁₀H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD07808559
CAS Number: 25594-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR