N-Methyl-1-quinolin-6-ylmethanamine 95%

Supplier: Apollo Scientific
Danger

Synonyms: 6-[(Methylamino)methyl]quinoline

OR9670-1G OR9670-250MG
APOSOR9670-1GEA 234 EUR
APOSOR9670-1G APOSOR9670-250MG
N-Methyl-1-quinolin-6-ylmethanamine 95%
N-Methyl-1-quinolin-6-ylmethanamine
Formula: C₁₁H₁₂N₂
MW: 172,23 g/mol
Boiling Pt: 120 °C/0,525mmHg
Storage Temperature: Ambient
MDL Number: MFCD06738897
CAS Number: 179873-36-0
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR