(4-Methylthien-2-yl)methylamine 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-(Aminomethyl)-4-methylthiophene
OR9699-1G
OR9699-250MG
APOSOR9699-1GEA
315
EUR
APOSOR9699-1G
APOSOR9699-250MG
(4-Methylthien-2-yl)methylamine 97%
(4-Methylthien-2-yl)methylamine
Formula:
C₆H₉NS MW: 127,21 g/mol Boiling Pt: 42…46 °C/0,1mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD06657973 CAS Number: 104163-39-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...