4-Pyrid-4-ylbenzoic acid 96%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Pyridin-4-yl-benzoic acid

OR9724-1G OR9724-250MG
APOSOR9724-1GEA 30.6 EUR
APOSOR9724-1G APOSOR9724-250MG
4-Pyrid-4-ylbenzoic acid 96%
4-Pyrid-4-ylbenzoic acid
Formula: C₁₂H₉NO₂
MW: 199,2100 g/mol
Melting Pt: 300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04114574
CAS Number: 4385-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR