4-Bromo-2-chloropyridine 98%
Supplier: Apollo Scientific
|
Warning
|
OR9753-25G
OR9753-5G
OR9753-100G
APOSOR9753-25GEA
30.6
EUR
APOSOR9753-25G
APOSOR9753-5G
APOSOR9753-100G
4-Bromo-2-chloropyridine 98%
4-Bromo-2-chloropyridine
Formula:
C₅H₃BrClN MW: 192.44 g/mol Melting Pt: 20…20.6 °C Density: 1.734 g/cm³ (25 °C) Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840756 CAS Number: 73583-37-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...