2-Oxo-4-phenylbutyric acid 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Oxo-4-phenylbutanoic acid
OR9775-100G
OR9775-25G
OR9775-5G
APOSOR9775-100GEA
616
EUR
APOSOR9775-100G
APOSOR9775-25G
APOSOR9775-5G
2-Oxo-4-phenylbutyric acid 97%
2-Oxo-4-phenylbutyric acid
Formula:
C₁₀H₁₀O₃ MW: 178,19 g/mol Melting Pt: 49 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00969471 CAS Number: 710-11-2 EINECS: 211-916-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...