Phthalazine 98%

Supplier: Apollo Scientific
Warning

OR9815-100G OR9815-25G OR9815-5G
APOSOR9815-100GEA 299 EUR
APOSOR9815-100G APOSOR9815-25G APOSOR9815-5G
Phthalazine 98%
Phthalazine
Formula: C₈H₆N₂
Boiling Pt: 315…317 °C/760mmHg
Melting Pt: 89…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006908
CAS Number: 253-52-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR