2-Amino-1-(4-chlorophenyl)-1-ethanol 95%

Supplier: Apollo Scientific
Warning

OR9825-1G OR9825-500MG
APOSOR9825-1GEA 314 EUR
APOSOR9825-1G APOSOR9825-500MG
2-Amino-1-(4-chlorophenyl)-1-ethanol 95%
2-Amino-1-(4-chlorophenyl)-1-ethanol
Formula: C₈H₁₀ClNO
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD06797373
CAS Number: 41870-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR