Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate

OR9842-1G OR9842-5G OR9842-10G
APOSOR9842-1GEA 21.9 EUR
APOSOR9842-1G APOSOR9842-5G APOSOR9842-10G
Ethyl-6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Ethyl 6-chloroimidazo[1,2-a]pyridine-2-carboxylate
Formula: C₁₀H₉ClN₂O₂
MW: 224,6437 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02186220
CAS Number: 67625-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR