2-(Phenylsulphonyl)aniline

Supplier: Apollo Scientific
Warning

OR9876-250G OR9876-25G OR9876-100G
APOSOR9876-250GEA 114 EUR
APOSOR9876-250G APOSOR9876-25G APOSOR9876-100G
2-(Phenylsulphonyl)aniline
2-(Phenylsulphonyl)aniline
Formula: C₁₂H₁₁NO₂S
MW: 233,2862 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00007706
CAS Number: 4273-98-7
EINECS: 224-271-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR