5-Methoxy-1-indanone 98%

Supplier: Apollo Scientific
Warning

OR9975-25G OR9975-5G OR9975-100G
APOSOR9975-25GEA 66.7 EUR
APOSOR9975-25G APOSOR9975-5G APOSOR9975-100G
5-Methoxy-1-indanone 98%
5-Methoxy-1-indanone
Formula: C₁₀H₁₀O₂
Melting Pt: 107…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00003789
CAS Number: 5111-70-6
EINECS: 225-838-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR