3-(3-(Trifluoromethoxy)phenyl)propionic acid 95%

Supplier: Apollo Scientific
Warning

PC0014-25G PC0014-5G PC0014-1G
APOSPC0014-25GEA 361 EUR
APOSPC0014-25G APOSPC0014-5G APOSPC0014-1G
3-(3-(Trifluoromethoxy)phenyl)propionic acid 95%
3-(3-(Trifluoromethoxy)phenyl)propionic acid
Formula: C₁₀H₉F₃O₃
MW: 234,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317834
CAS Number: 168833-77-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR