2-Bromo-4,6-difluoroanisole 99%
Supplier: Apollo Scientific
|
Warning
|
PC0038-100G
PC0038-25G
PC0038-500G
APOSPC0038-100GEA
116
EUR
APOSPC0038-100G
APOSPC0038-25G
APOSPC0038-500G
2-Bromo-4,6-difluoroanisole 99%
2-Bromo-4,6-difluoroanisole
Formula:
C₇H₅BrF₂O MW: 223,02 g/mol Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00070754 CAS Number: 202865-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...