2-Chloro-5-fluorobenzyl alcohol 97%
Supplier: Apollo Scientific
|
Warning
|
PC0057-1G
PC0057-5G
PC0057-25G
APOSPC0057-1GEA
21.9
EUR
APOSPC0057-1G
APOSPC0057-5G
APOSPC0057-25G
2-Chloro-5-fluorobenzyl alcohol 97%
2-Chloro-5-fluorobenzyl alcohol
Formula:
C₇H₆ClFO MW: 160,58 g/mol Melting Pt: 68…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631418 CAS Number: 261762-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...