2-Bromo-6-fluorotoluene 98%
Supplier: Apollo Scientific
|
Warning
|
PC0066-100G
PC0066-25G
PC0066-500G
APOSPC0066-100GEA
81.2
EUR
APOSPC0066-100G
APOSPC0066-25G
APOSPC0066-500G
2-Bromo-6-fluorotoluene 98%
2-Bromo-6-fluorotoluene
Formula:
C₇H₆BrF MW: 189,03 g/mol Boiling Pt: 180 °C Density: 1,53 g/cm³ (25 °C) Flash Pt: 76 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051438 CAS Number: 1422-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...