3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde 98%

Supplier: Apollo Scientific
Warning

PC0144-1G PC0144-25G PC0144-5G
APOSPC0144-1GEA 21.9 EUR
APOSPC0144-1G APOSPC0144-25G APOSPC0144-5G
3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde 98%
3-Chloro-2-fluoro-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₃ClF₄O
Boiling Pt: 196 °C
Density: 1,54 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD01631510
CAS Number: 261763-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR