2-Fluoro-4-pyridone 95%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoropyridin-4-ol, 2-Fluoro-4-hydroxypyridine

PC01475-1G PC01475-5G
APOSPC01475-1GEA 114 EUR
APOSPC01475-1G APOSPC01475-5G
2-Fluoro-4-pyridone 95%
2-Fluoro-4-pyridone
Formula: C₅H₄FNO
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD04114159
CAS Number: 22282-69-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR