3-Chloro-2-fluoro-6-(trifluoromethyl)benzamide 97%

Supplier: Apollo Scientific
Warning

PC0149-5G PC0149-1G
APOSPC0149-5GEA 306 EUR
APOSPC0149-5G APOSPC0149-1G
3-Chloro-2-fluoro-6-(trifluoromethyl)benzamide 97%
3-Chloro-2-fluoro-6-(trifluoromethyl)benzamide
Formula: C₈H₄ClF₄NO
Storage Temperature: Ambient
MDL Number: MFCD01631400
CAS Number: 186517-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR