4-Chloro-2-fluorobenzonitrile 98%
Supplier: Apollo Scientific
|
Danger
|
PC0162-1G
PC0162-25G
PC0162-5G
APOSPC0162-1GEA
14.7
EUR
APOSPC0162-1G
APOSPC0162-25G
APOSPC0162-5G
4-Chloro-2-fluorobenzonitrile 98%
4-Chloro-2-fluorobenzonitrile
Formula:
C₇H₃ClFN MW: 155,56 g/mol Boiling Pt: 92 °C/22mmHg Melting Pt: 58…61 °C Storage Temperature: Ambient |
MDL Number:
MFCD00143284 CAS Number: 57381-51-8 EINECS: 260-712-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...