4-Chloro-3-(trifluoromethyl)benzyl bromide 98%

Supplier: Apollo Scientific
Danger

Synonyms: 1-(Bromomethyl)-4-chloro-3-(trifluoromethyl)benzene, 4-(Bromomethyl)-1-chloro-2-(trifluoromethyl)benzene

PC0194-100G PC0194-25G PC0194-5G
APOSPC0194-100GEA 213 EUR
APOSPC0194-100G APOSPC0194-25G APOSPC0194-5G
4-Chloro-3-(trifluoromethyl)benzyl bromide 98%
4-Chloro-3-(trifluoromethyl)benzyl bromide
Formula: C₈H₅BrClF₃
MW: 273,48 g/mol
Melting Pt: 36…40 °C
Storage Temperature: Ambient
MDL Number: MFCD01631540
CAS Number: 261763-23-9
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR