(2,4-Dichloro-5-fluorophenyl)acetonitrile

Supplier: Apollo Scientific
Danger

Synonyms: 2-(2,4-Dichloro-5-fluorophenyl)acetonitrile

PC0217-1G PC0217-5G
APOSPC0217-1GEA 118 EUR
APOSPC0217-1G APOSPC0217-5G
(2,4-Dichloro-5-fluorophenyl)acetonitrile
(2,4-Dichloro-5-fluorophenyl)acetonitrile
Formula: C₈H₄Cl₂FN
MW: 204,300 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631382
CAS Number: 261763-28-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR