3-Fluoro-4-methylcinnamic acid 98%

Supplier: Apollo Scientific

Synonyms: 3-(3-Fluoro-4-methylphenyl)acrylic acid

PC0366-10G PC0366-5G PC0366-1G
APOSPC0366-10GEA 249 EUR
APOSPC0366-10G APOSPC0366-5G APOSPC0366-1G
3-Fluoro-4-methylcinnamic acid 98%
3-Fluoro-4-methylcinnamic acid
Formula: C₁₀H₉FO₂
MW: 180,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631413
CAS Number: 261951-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR