2-Fluoro-6-methoxybenzoic acid 98%
Supplier: Apollo Scientific
|
Warning
|
PC0412-5G
PC0412-100G
PC0412-25G
APOSPC0412-5GEA
24.8
EUR
APOSPC0412-5G
APOSPC0412-100G
APOSPC0412-25G
2-Fluoro-6-methoxybenzoic acid 98%
2-Fluoro-6-methoxybenzoic acid
Formula:
C₈H₇FO₃ Melting Pt: 85…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00671764 CAS Number: 137654-21-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...