Tris(trifluoro-2,4-pentanedionato)aluminum(III) 98%

Supplier: Apollo Scientific

Synonyms: Aluminum(III) Trifluoroacetylacetonate, Trifluoroacetylacetonato Aluminum(III) Salt

PC0429-25G PC0429-5G
APOSPC0429-25GEA 945 EUR
APOSPC0429-25G APOSPC0429-5G
Tris(trifluoro-2,4-pentanedionato)aluminum(III) 98%
Tris(trifluoro-2,4-pentanedionato)aluminum(III)
Formula: C₁₅H₁₂AlF₉O₆
MW: 486,22 g/mol
Melting Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00042062
CAS Number: 14354-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR