5-Amino-2-methylbenzotrifluoride 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Methyl-3-(trifluoromethyl)aniline

PC0440-25G PC0440-5G PC0440-100G
APOSPC0440-25GEA 77.1 EUR
APOSPC0440-25G APOSPC0440-5G APOSPC0440-100G
5-Amino-2-methylbenzotrifluoride 98%
5-Amino-2-methylbenzotrifluoride
Formula: C₈H₈F₃N
MW: 175,16 g/mol
Boiling Pt: 204 °C
Density: 1,22 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Ambient
MDL Number: MFCD01631582
CAS Number: 65934-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR