2-Methyl-3-(trifluoromethyl)benzoic acid 97%

Supplier: Apollo Scientific
Warning

PC0446-1G PC0446-25G PC0446-5G
APOSPC0446-1GEA 20.5 EUR
APOSPC0446-1G APOSPC0446-25G APOSPC0446-5G
2-Methyl-3-(trifluoromethyl)benzoic acid 97%
2-Methyl-3-(trifluoromethyl)benzoic acid
Formula: C₉H₇F₃O₂
MW: 204,15 g/mol
Melting Pt: 106…107 °C
Storage Temperature: Ambient
MDL Number: MFCD01631588
CAS Number: 62089-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR