4-Methyl-2-(trifluoromethyl)benzoic acid
Supplier: Apollo Scientific
|
Warning
|
PC0449-5G
PC0449-10G
PC0449-1G
PC0449-25G
APOSPC0449-5GEA
101
EUR
APOSPC0449-5G
APOSPC0449-10G
APOSPC0449-1G
APOSPC0449-25G
4-Methyl-2-(trifluoromethyl)benzoic acid
4-Methyl-2-(trifluoromethyl)benzoic acid
Formula:
C₉H₇F₃O₂ MW: 204,15 g/mol Melting Pt: 135…141 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631591 CAS Number: 120985-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...