4-Methyl-3-(trifluoromethyl)benzoic acid
Supplier: Apollo Scientific
|
Warning
|
PC0450-25G
PC0450-5G
APOSPC0450-25GEA
82.9
EUR
APOSPC0450-25G
APOSPC0450-5G
4-Methyl-3-(trifluoromethyl)benzoic acid
4-Methyl-3-(trifluoromethyl)benzoic acid
Formula:
C₉H₇F₃O₂ MW: 204,15 g/mol Melting Pt: 175…178 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631592 CAS Number: 261952-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...