1-(4-Fluoro-2-nitrophenyl)pyrrolidine
Supplier: Apollo Scientific
|
Warning
|
PC0452-1G
PC0452-5G
APOSPC0452-1GEA
14.7
EUR
APOSPC0452-1G
APOSPC0452-5G
1-(4-Fluoro-2-nitrophenyl)pyrrolidine
1-(4-Fluoro-2-nitrophenyl)pyrrolidine
Formula:
C₁₀H₁₁ClN₂O₂ MW: 210,20 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00798372 CAS Number: 778-56-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...