2-Methyl-3-(trifluoromethyl)benzoyl chloride

Supplier: Apollo Scientific
Danger

PC0464-10G PC0464-5G
APOSPC0464-10GEA 328 EUR
APOSPC0464-10G APOSPC0464-5G
2-Methyl-3-(trifluoromethyl)benzoyl chloride
2-Methyl-3-(trifluoromethyl)benzoyl chloride
Formula: C₉H₆ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD01631598
CAS Number: 261952-07-2
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR