5-Iodooctafluoro-3-oxapentanesulphonyl fluoride

Supplier: Apollo Scientific
Danger

Synonyms: Tetrafluoro-2-(tetrafluoro-2-iodoethoxy)ethanesulphonyl Fluoride

PC0471-25G
APOSPC0471-25GEA 168 EUR
APOSPC0471-25G
5-Iodooctafluoro-3-oxapentanesulphonyl fluoride
5-Iodooctafluoro-3-oxapentanesulphonyl fluoride
Formula: C₄F₉IO₃S
MW: 426 g/mol
Boiling Pt: 126…127 °C
Density: 2,051 g/cm³ (25 °C)
Flash Pt: >100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798139
CAS Number: 66137-74-4
EINECS: 266-172-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR