5-Bromo-2-methylbenzotrifluoride 98%

Supplier: Apollo Scientific
Warning

PC0491-1G PC0491-25G PC0491-5G
APOSPC0491-1GEA 21.9 EUR
APOSPC0491-1G APOSPC0491-25G APOSPC0491-5G
5-Bromo-2-methylbenzotrifluoride 98%
5-Bromo-2-methylbenzotrifluoride
Formula: C₈H₆BrF₃
Flash Pt: 78 °C
Storage Temperature: Ambient
MDL Number: MFCD01631587
CAS Number: 86845-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR