2-Amino-5-chloro-2'-fluorobenzophenone
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
(2-Amino-5-chlorophenyl)(2-fluorophenyl)methanone, 2-Amino-5-chloro-2′-fluorobenzophenone
PC0492-100G
APOSPC0492-100GEA
66.7
EUR
APOSPC0492-100G
2-Amino-5-chloro-2'-fluorobenzophenone
2-Amino-5-chloro-2'-fluorobenzophenone
Formula:
C₁₃H₉ClFNO MW: 249,67 g/mol Melting Pt: 95…98 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00038381 CAS Number: 784-38-3 EINECS: 212-316-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...